missing translation for 'onlineSavingsMsg'
Learn More
SCH 39166 hydrobromide, Tocris Bioscience™ Product Code.: 18004112

SCH 39166 hydrobromide, Tocris Bioscience™

Product Code. 18004112
50 mg
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Product Code. 18004112

Brand: Tocris Bioscience 2299/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High affinity D1-like antagonist

High affinity dopamine D1/D5 receptor antagonist; displays Ki values of 1.2, 2, 980, 5520, 80 and 731 nM for binding to D1, D5, D2, D4, 5-HT and α2a receptors, respectively.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1227675-51-5
Molecular Formula C19H21BrClNO
Molecular Weight (g/mol) 394.737
InChI Key GAUWIDFICGEZKR-UHFFFAOYSA-N
Synonym (6aS-trans)-11-Chloro-6,6a,7,8,9,13b-hexahydro-7-methyl-5H-benzo[d]naphth[2,1-b]azepin-12-ol hydrobromide
PubChem CID 73324728
IUPAC Name 11-chloro-7-methyl-5,6,6a,8,9,13b-hexahydronaphtho[1,2-a][3]benzazepin-12-ol;hydrobromide
SMILES CN1CCC2=CC(=C(C=C2C3C1CCC4=CC=CC=C34)O)Cl.Br

Specifications

Quantity 50 mg
Formula Weight 394.73
Percent Purity >99%
Chemical Name or Material SCH 39166 hydrobromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
SCH 39166 hydrobromide, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.