Learn More
Sartorius Replacement Part for Polycarbonate Filtration Holders
Replacement Part for Polycarbonate Filtration Holders is for use with Microsart™ Manifolds 16510 and 16511.
10.50€ - 125.00€
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Quantity | Price | Quantity & Availability | |||||
10079740
|
Sartorius
6980225 |
Plug | 10 Pk. |
17.05€
Pack of 10 |
In Stock
Log in to see stock. |
|||||
10355461
|
Sartorius
6980227 |
Filter System Stopper | 1 Ea. |
10.50€
Each |
In Stock
Log in to see stock. |
|||||
15972630
|
Sartorius
6980230 |
Top Part | 1 Ea. |
29.65€
Each |
In Stock
Log in to see stock. |
|||||
15982630
|
Sartorius
6980232 |
Filter Support | 2 Pk. |
14.80€
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
15992630
|
Sartorius
6980234 |
Hose Nipple | 1 Ea. |
15.75€
Each |
In Stock
Log in to see stock. |
|||||
15902640
|
Sartorius
6980235 |
Silicone O-ring | 3 Pk. |
125.00€
Each |
In Stock
Log in to see stock. ![]() |
|||||
15912660
|
Sartorius
6981090 |
Receiver Flask | 1 Ea. |
36.20€
Each |
In Stock
Log in to see stock. |
|||||
Description
- Polycarbonate Accessory
- For use with Microsart™ Manifolds 16510, 16511
Specifications
Reusable | |
Non-sterile |
Product Title
Sartorius Replacement Part for Polycarbonate Filtration Holders