Learn More
Thermo Scientific Chemicals Sapphire substrate, 50.8mm dia x 0.432mm thick, EPI polished one side, C-axis, HEMCOR single crystal
Brand: Thermo Scientific Chemicals 045019.KU
1107.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Sapphire is used as an insulating substrate upon which to deposit silicon to make the integrated circuits, high-power radio-frequency (RF) applications such as those found in cellular telephones, public-safety band radios, and satellite communication systems, used in the semiconductor industry, blue light-emitting diodes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sapphire Substrate, HEMCOR Single Crystal | |
50.8 mm | |
50.8mm dia. x 0.432 mm thick |
EPI polished one side | |
5 Ea. | |
0.432 mm |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
U09K038 | Certificate of Analysis | 14/08/2024 | 045019.KU,045019.KT |
S05J034 | Certificate of Analysis | 13/06/2023 | 045019.KU |
J15S031 | Certificate of Analysis | 08/12/2021 | 045019.KU,045019.KT,045020.KT |
Q23H049 | Certificate of Analysis | 07/12/2021 | 045019.KU,045019.KT,045020.KT |
N13A028 | Certificate of Analysis | 07/12/2021 | 045019.KU,045019.KT,045020.KT |