Learn More
Tocris Bioscience™ SAG
Potent Smoothened receptor agonist; activates the Hedgehog signaling pathway
232.00€ - 820.00€
Specifications
Purity | >99% |
---|---|
Product Type | SAG |
Molecular Weight (g/mol) | 490.06 |
Inhibitors | SAG |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18024052
|
Tocris Bioscience™
4366/10 |
10 mg |
820.00€
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18709807
|
Tocris Bioscience™
4366/1 |
1 mg |
232.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SAG is a potent Smoothened (Smo) receptor agonist (Kd = 59 nM); antagonizes Cyclopamine (Cat. No. 1623) action at the Smo receptor. SAG potently activates the Hedgehog signaling pathway in Shh-light 2 cells (EC50 ∼ 3 nM) and nduces pathway activation independently of Ptch proteins. SAG is a putative inhibitor of a cellular component required for Hedgehog signaling and also enhances neuronal differentiation of iPSCs into dopaminergic neurons.SAG dihydrochloride (Cat. No. 6390) also available.
Specifications
>99% | |
SAG | |
490.06 | |
SAG |
For Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.