Learn More
Tocris Bioscience™ SAG
Potent Smoothened receptor agonist; activates the Hedgehog signaling pathway
Brand:  Tocris Bioscience™ 4366/1
 View more versions of this product
Product Code. 18709807
  • 232.00€ / 1mg
Explore more special offers
Description
Description
SAG is a potent Smoothened (Smo) receptor agonist (Kd = 59 nM); antagonizes Cyclopamine (Cat. No. 1623) action at the Smo receptor. SAG potently activates the Hedgehog signaling pathway in Shh-light 2 cells (EC50 ∼ 3 nM) and nduces pathway activation independently of Ptch proteins. SAG is a putative inhibitor of a cellular component required for Hedgehog signaling and also enhances neuronal differentiation of iPSCs into dopaminergic neurons.
SAG dihydrochloride (Cat. No. 6390) also available.
Specifications
Specifications
>99%
SAG
SAG
1 mg
490.06