missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Safety Cover for Heat-On Block

This cover for Heat-On Blocks provides an extra level of protection against burns when heating round bottom flasks.

554.00€ - 954.00€

Specifications

Color White
For Use With (Application) Heat Protection
Material PTFE
View More Specs

Products 3
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
13400049
View Documents
Heidolph™
505-80081-00
200 to 300 mL Round Bottom Flask
554.00€
Each
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
 
12397353
View Documents
Heidolph™
505-80082-00
500 mL Round Bottom Flask
585.00€
Each
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
 
11367080
View Documents
Heidolph™
505-80083-00
1 L Round Bottom Flask
954.00€
Each
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
 
Description

Description

  • Ensures more protection against burns when using Heat-On Block
  • Despite reduced danger when using Heat-On Blocks as compared to conventional heating bath, risk of burns is not zero- this safety cover reduces that
  • Cover is made of chemical-resistant, easy-to-clean, PTFE
Specifications

Specifications

White
Heat Protection
PTFE
Videos
SDS
Documents

Documents

Product Certifications