Learn More
Heidolph™ Safety Cover for Heat-On Block
This cover for Heat-On Blocks provides an extra level of protection against burns when heating round bottom flasks.
Brand:  Heidolph™ 505-80082-00
 View more versions of this product
Product Code. 12397353
  • 585.00€ / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Description
  • Ensures more protection against burns when using Heat-On Block
  • Despite reduced danger when using Heat-On Blocks as compared to conventional heating bath, risk of burns is not zero- this safety cover reduces that
  • Cover is made of chemical-resistant, easy-to-clean, PTFE
Specifications
Specifications
White
500 mL Round Bottom Flask
Heat Protection
PTFE