missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Selective σ1 receptor agonist (IC50 = 17.4 nM). Exhibits selectivity for σ1 over σ2 receptors. Inhibits angiotensin II-induced cardiomyocyte hypertrophy in vitro and attenuates myocardial hypertrophy in vivo. Enhances brain plasticity and sensorimotor function in a rat model of experimental stroke.

Specifications
Specifications
Color | White |
Quantity | 50 mg |
Synonym | 1-3,4-dimethoxyphenethyl-4-3-phenylpropyl piperazine dihydrochloride, sa4503 dihydrochloride, sa 4503 dihydrochloride, 1-2-3,4-dimethoxyphenyl ethyl-4-3-phenylpropyl piperazine dihydrochloride, 1-3,4-dimethoxyphenethyl-4-3-phenylpropyl piperazine 2hcl, sa-4503 dihydrochloride, 1-2-3,4-dimethoxyphenyl ethyl-4-3-phenylpropyl-piperazine dihydrochloride, 1-2-3,4-dimethoxyphenyl ethyl-4-3-phenylpropyl piperazine-hydrogen chloride 1/2 |
Formula Weight | 441.43 |
Percent Purity | >98% |
Physical Form | Solid |
Chemical Name or Material | SA 4503 dihydrochloride |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 10
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
SA 4503 dihydrochloride, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction