missing translation for 'onlineSavingsMsg'
Learn More
(S)-(-)-Propranolol hydrochloride, Tocris Bioscience™ Product Code.: 18015392

(S)-(-)-Propranolol hydrochloride, Tocris Bioscience™

Product Code. 18015392
100 mg
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy

Product Code. 18015392

Brand: Tocris Bioscience 0834/100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

β antagonist. More active enantiomer of propranolol (Cat. No. 0624)

More active enantiomer of the β-adrenoceptor antagonist propranolol .

Racemate, R-enantiomer and Glycol Metabolite also available.

TRUSTED_SUSTAINABILITY

Specifications

CAS 4221-03-8
Molecular Formula C5H10O2
Quantity 100 mg
Synonym pentanal, 5-hydroxy, 5-hydroxy-pentanal, 5-?hydroxypentanal, 5-hydroxyvaleraldehyde, cas
InChI Key CNRGMQRNYAIBTN-UHFFFAOYSA-N
SMILES C(CCO)CC=O
IUPAC Name 5-hydroxypentanal
Molecular Weight (g/mol) 102.13
PubChem CID 20205
Formula Weight 295.81
Percent Purity >98%
Chemical Name or Material (S)-(-)-Propranolol hydrochloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-(-)-Propranolol hydrochloride, Tocris Bioscience™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.