Learn More
(S)-(+)-2-Aminobutyramide hydrochloride, 99%, Thermo Scientific Chemicals
26.10€ - 209.00€
Chemical Identifiers
CAS | 7682-20-4 |
---|---|
Molecular Formula | C4H11ClN2O |
Molecular Weight (g/mol) | 138.595 |
MDL Number | MFCD00136565 |
InChI Key | HDBMIDJFXOYCGK-DFWYDOINSA-N |
Synonym | s-2-aminobutanamide hydrochloride, s-2-aminobutyramide hydrochloride, h-abu-nh2 hcl, 2s-2-aminobutanamide hydrochloride, l-2-aminobutanamide hydrochloride, l-homoalanylamide hcl, s-+-2-aminobutanamide hydrochloride, butanamide, 2-amino-, monohydrochloride, 2s, s-2-aminobutanamide hcl, s-2-amino butanamide hcl |
PubChem CID | 11961924 |
IUPAC Name | (2S)-2-aminobutanamide;hydrochloride |
SMILES | CCC(C(=O)N)N.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15481878
|
Thermo Scientific Alfa Aesar
H55576.MD |
250 mg |
26.10€
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15461878
|
Thermo Scientific Alfa Aesar
H55576.03 |
1 g |
59.50€
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15471878
|
Thermo Scientific Alfa Aesar
H55576.06 |
5 g |
209.00€
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7682-20-4 | |
138.595 | |
HDBMIDJFXOYCGK-DFWYDOINSA-N | |
11961924 | |
CCC(C(=O)N)N.Cl |
C4H11ClN2O | |
MFCD00136565 | |
s-2-aminobutanamide hydrochloride, s-2-aminobutyramide hydrochloride, h-abu-nh2 hcl, 2s-2-aminobutanamide hydrochloride, l-2-aminobutanamide hydrochloride, l-homoalanylamide hcl, s-+-2-aminobutanamide hydrochloride, butanamide, 2-amino-, monohydrochloride, 2s, s-2-aminobutanamide hcl, s-2-amino butanamide hcl | |
(2S)-2-aminobutanamide;hydrochloride |
Specifications
7682-20-4 | |
C4H11ClN2O | |
s-2-aminobutanamide hydrochloride, s-2-aminobutyramide hydrochloride, h-abu-nh2 hcl, 2s-2-aminobutanamide hydrochloride, l-2-aminobutanamide hydrochloride, l-homoalanylamide hcl, s-+-2-aminobutanamide hydrochloride, butanamide, 2-amino-, monohydrochloride, 2s, s-2-aminobutanamide hcl, s-2-amino butanamide hcl | |
+24° (c=1 in Water) | |
(2S)-2-aminobutanamide;hydrochloride | |
11961924 | |
99% |
259°C to 263°C | |
MFCD00136565 | |
HDBMIDJFXOYCGK-DFWYDOINSA-N | |
CCC(C(=O)N)N.Cl | |
138.595 | |
138.6 | |
(S)-(+)-2-Aminobutyramide hydrochloride |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only