missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Aminobutyramide hydrochloride, 99%, Thermo Scientific Chemicals

Product Code. 15461878
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7682-20-4
Molecular Formula C4H11ClN2O
Molecular Weight (g/mol) 138.595
MDL Number MFCD00136565
InChI Key HDBMIDJFXOYCGK-DFWYDOINSA-N
Synonym s-2-aminobutanamide hydrochloride, s-2-aminobutyramide hydrochloride, h-abu-nh2 hcl, 2s-2-aminobutanamide hydrochloride, l-2-aminobutanamide hydrochloride, l-homoalanylamide hcl, s-+-2-aminobutanamide hydrochloride, butanamide, 2-amino-, monohydrochloride, 2s, s-2-aminobutanamide hcl, s-2-amino butanamide hcl
PubChem CID 11961924
IUPAC Name (2S)-2-aminobutanamide;hydrochloride
SMILES CCC(C(=O)N)N.Cl

Specifications

Melting Point 259°C to 263°C
Quantity 1 g
Optical Rotation +24° (c=1 in Water)
Formula Weight 138.6
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-Aminobutyramide hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'