Learn More
Bollé Safety Rush+ Platinum™ Twilight Safety Glasses Product Code.: 15226705
Add to basket
Bollé Safety Rush+ Platinum™ Twilight Safety Glasses
Product Code. 15226705
Product Code. 15226705
Brand: Bollé Safety RUSHPTWI
Edge
Add to basket
Add to basket
The RUSH + adopts an ultra-sporty design with its co-injected and ultra-flexible temples
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog, Scratch-resistant
Product Type Rush RUSHPTWI Safety Spectacle
No. per Pack 1 Ea.
  • Bollé Safety - Rush+
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Bolle Safety Rush+
    • Bollé Safety - Platinum coating
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Bolle Safety Platinum coating
      • Bollé Safety - Platinum efficiency demonstration
        Video Player is loading.
        Current Time 0:00
        Duration 0:00
        Loaded: 0%
        Stream Type LIVE
        Remaining Time 0:00
        1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
          Bolle Safety Platinum efficiency demonstration
        • Multiple Manufacturers
          Save up to 60% on gloves, apparel, and more
          Promo Code: 24131
          Expires2025-06-08
          New Offer
          Shop Now
        Product Content Correction
        Product Title
        Bollé Safety Rush+ Platinum™ Twilight Safety Glasses >