missing translation for 'onlineSavingsMsg'
Learn More

RPS29 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

145.00€ - 410.00€

Specifications

Antigen RPS29
Concentration 0.13 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16867254
View Documents
Proteintech
17374-1-AP-20UL
20 μL
145.00€
20µL
Please to purchase this item. Need a web account? Register with us today!
16857254
View Documents
Proteintech
17374-1-AP-150UL
150 μL
410.00€
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPS29 is a ribosomal protein that is a component of the 40S subunit and a member of the S14P family of ribosomal proteins. The protein, which contains a C2-C2 zinc finger-like domain that can bind to zinc, can enhance the tumor suppressor activity of Ras-related protein 1A (KREV1). It is located in the cytoplasm.
Specifications

Specifications

RPS29
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human
P62273
6235
RPS29 Fusion Protein Ag11179
Primary
-20°C
RPS29
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
40S ribosomal protein S29, ribosomal protein S29, RPS29
RPS29
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications