Learn More
RPS29 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17374-1-AP-20UL
 View more versions of this product
Product Code. 16867254
  • 145.00€ / 20µL
Explore more special offers
Description
Description
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. RPS29 is a ribosomal protein that is a component of the 40S subunit and a member of the S14P family of ribosomal proteins. The protein, which contains a C2-C2 zinc finger-like domain that can bind to zinc, can enhance the tumor suppressor activity of Ras-related protein 1A (KREV1). It is located in the cytoplasm.
Specifications
Specifications
RPS29
Polyclonal
Unconjugated
RPS29
40S ribosomal protein S29, ribosomal protein S29, RPS29
Rabbit
Antigen Affinity Chromatography
RUO
6235
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62273
RPS29
RPS29 Fusion Protein Ag11179