Learn More
Rotronic™ Disposable Sample Cups
Ensures optimum sample volume is introduced into the sample holders.
229.00€ - 251.00€
Specifications
Product Type | Sample Cup |
---|---|
Disposable | Single-use |
For Use With (Application) | Collecting and Storing samples |
Product Code | Brand | Depth (English) | Depth (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Depth (English) | Depth (Metric) | For Use With (Equipment) | Price | Quantity & Availability | ||||
11950685
|
Rotronic™
PS14 |
0.55 in. | 1.4 cm | WP-14-S sample holders |
229.00€
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11940685
|
Rotronic™
PS40 |
1.57 in. | 4 cm | WP-40 or WP-40TH sample holders |
251.00€
Pack of 100 |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||
Description
- Prevent sample holders from coming into direct contact with product being tested, thereby preventing soiling or cross contamination
- Provides convenient means of collecting and storing samples
Specifications
Sample Cup | |
Single-use | |
Collecting and Storing samples |
Product Title
Rotronic™ Disposable Sample Cups