Learn More
IKA™ ROTAVISC Standard Spindle
Standard spindle designed for use with IKA™ ROTAVISC Viscometers.
48.00€ - 94.50€
Specifications
Product Type Standard Spindle
For Use With (Equipment) IKA™ ROTAVISC Viscometer
Material AISI 316L
View More Specs
Products 10
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
17277539
View Documents
IKA™
0025006828
SP-1 Standard Spindle
94.50€
Each
Estimated Shipment: 11-07-2025
to see stock.
Add to basket
17287539
View Documents
IKA™
0025006829
SP-2 Standard Spindle
89.50€
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17297539
View Documents
IKA™
0025006830
SP-3 Standard Spindle
87.00€
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17207549
View Documents
IKA™
0025006831
SP-4 Standard Spindle
81.50€
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17217549
View Documents
IKA™
0025006834
SP-7 Standard Spindle
68.50€
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17227549
View Documents
IKA™
0025006835
SP-8 Standard Spindle
66.50€
Each
Estimated Shipment: 13-06-2025
to see stock.
Add to basket
17237549
View Documents
IKA™
0025006836
SP-9 Standard Spindle
66.50€
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17247549
View Documents
IKA™
0025006837
SP-10 Standard Spindle
63.00€
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17257549
View Documents
IKA™
0025006838
SP-11 Standard Spindle
60.00€
Each
Estimated Shipment: 10-06-2025
to see stock.
Add to basket
17267549
View Documents
IKA™
0025006839
SP-12 Standard Spindle
48.00€
Each
In Stock
to see stock.
Add to basket
Description
Description
  • Quick coupling
  • Hook hitch
  • M2.5 left-handed thread
Specifications
Specifications
Standard Spindle
IKA™ ROTAVISC Viscometer
AISI 316L
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
IKA™ ROTAVISC Standard Spindle