missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann™ Rotarus™ PKF 60-16-3

High quality PKF 60-series single-channel pump head that can be used in combination with stopper-less tubes.

Brand:  Hirschmann™ 951106060

Product Code. 16497039

  • 470.00€ / Each
Estimated Shipment: 17-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • 3 rollers
  • For WT 1.6 mm tubes
  • Only an adapter plate is necessary for the connection between pump head and pump drive
  • The pump head of the flow and volume pump drives is recognized by RFID technology
  • A simple and easy loading of the tubing is given by unlocking the pump head
  • The pump head is made of durable PPS, a high-performance plastic with very high heat resistance, high chemical resistance and durability
Specifications

Specifications

Approx. 5%
Black
Generally from 0.04 to 2150 mL/min. per channel
Polyphenylene Sulfide
106 mm
94 mm
Stainless Steel 304
PPS
0.2 to 500 L/min
54 x 94 x 106 mm
Pump drive and tubing
3
54 mm
1
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hirschmann™ Rotarus™ PKF 60-16-3 > Color: Black

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.