Learn More
Rogo Sampaic™ Glass Coverslips
Preserve and protect specimens on slides. Rogo Sampaic™ Glass Coverslips are manufactured from borosilicate glass for excellent transparency and prevention of color distortion.
Brand:  Rogo Sampaic™ BPD025
 View more versions of this product
Product Code. 11983564
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Explore more special offers
Description
Description
  • Available in a range of sizes
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Cover Slide
Borosilicate Glass
60 mm
Slide microscopy
100/Pk.
Rectangle
24 mm
24 x 60 mm
ISO 8255