Learn More
Rogo Sampaic™ Glass Coverslips
Preserve and protect specimens on slides. Rogo Sampaic™ Glass Coverslips are manufactured from borosilicate glass for excellent transparency and prevention of color distortion.
17.00€ - 75.40€
Specifications
Product Type | Cover Slide |
---|---|
Material | Borosilicate Glass |
For Use With (Application) | Slide microscopy |
Certifications | ISO 8255 |
Product Code | Brand | Length (Metric) | Width (Metric) | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Width (Metric) | Quantity | Price | Quantity & Availability | ||||||
11993564
|
Rogo Sampaic™
BPD016 |
18 mm | 18 mm | 1000/Pk. |
43.00€
Pack of 1000 |
In Stock
Log in to see stock. |
||||||
11707394
|
Rogo Sampaic™
BPD017 |
20 mm | 20 mm | 200/Pk. |
20.00€
Pack of 200 |
In Stock
Log in to see stock. ![]() |
||||||
11757394
|
Rogo Sampaic™
BPD024 |
24 mm | 50 mm | 100/Pk. |
20.80€
Pack of 100 |
In Stock
Log in to see stock. ![]() |
||||||
11717394
|
Rogo Sampaic™
BPD018 |
22 mm | 22 mm | 200/Pk. |
23.00€
Pack of 200 |
In Stock
Log in to see stock. |
||||||
11983564
|
Rogo Sampaic™
BPD025 |
24 mm | 60 mm | 100/Pk. | N/A | N/A | N/A | |||||
11767394
|
Rogo Sampaic™
BPD025 |
24 mm | 60 mm | 100/Pk. |
17.00€
Pack of 100 |
In Stock
Log in to see stock. |
||||||
Description
- Available in a range of sizes
- Corrosion-free and resistant to thermal shock
Specifications
Cover Slide | |
Borosilicate Glass | |
Slide microscopy | |
ISO 8255 |
Product Title
Rogo Sampaic™ Glass Coverslips