Learn More
SI Analytics™ Resistance Thermometer with Screw Plug Head
Features a glass shaft and Pt 100 sensor
Brand:  SI Analytics™ 285119162
Product Code. 10747301
  • 307.00€ / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Screw plug head
  • Type W 2081
  • Glass shaft
  • Sensor Pt 100
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12 mm
120 mm
-30°C to +135°C
with Screw Plug Head, Type W 2081, Sensor Pt 100
Resistance thermometer
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
SI Analytics™ Resistance Thermometer with Screw Plug Head > 12 mm