missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Reserpin-d9, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support

Specifications
Specifications
CAS | 84759-11-5 |
Packaging | Glass Bottle |
Quantity | 10 mg |
Molecular Formula | C33D9H31N2O9 |
MDL Number | MFCD23097077 |
For Use With (Application) | Alkaloids |
Physical Form | Powder |
Molecular Weight (g/mol) | 617.73 |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Reserpin-d9, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction