Learn More
Thermo Scientific™ Remel™ Reinforced Medium for Clostridia MLT (Dehydrated)
Remel Reinforced Medium for Clostridia MLT (Dehydrated) facilitates cultivation and enumeration of Clostridium species and other anaerobes.
Brand:  Thermo Scientific™ R455402
447.92 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 12697619
  • 460.00€ / Each
Explore more special offers
Description
Description
Remel Reinforced Medium for Clostridia MLT (Dehydrated) facilitates cultivation and enumeration of Clostridium species and other anaerobes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Industrial Reference: USP
Powder
500 g
Reinforced Medium for Clostridia MLT
Dehydrated Culture Media
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number814283 Certificate TypeCertificate of Analysis Date26/07/2024 Catalog NumberR455402
Lot Number653596 Certificate TypeCertificate of Analysis Date09/04/2024 Catalog NumberR455402
Lot Number277475 Certificate TypeCertificate of Analysis Date08/04/2024 Catalog NumberR455402
Lot Number463115 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog NumberR455402
Lot Number182508 Certificate TypeCertificate of Analysis Date03/04/2024 Catalog NumberR455402
Product Title
Thermo Scientific™ Remel™ Reinforced Medium for Clostridia MLT (Dehydrated) > 500g