Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent
Used for serological identification of E. coli by slide and/or tube agglutination tests.
Brand: Thermo Scientific™ R30954901
125.09 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Remel Agglutinating Sera, E. coli Polyvalent is used for serological identification of E. coli by slide and/or tube agglutination tests.
Specifications
E. coli Polyvalent 2 (types O26, O55, O111, O119, O126) | |
Agglutinating Sera | |
2 mL |
E. coli | |
2 mL/Vial |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3762992 | Certificate of Analysis | 29/01/2025 | R30954901 |
3660933 | Certificate of Analysis | 02/08/2023 | R30954901 |
3568068 | Certificate of Analysis | 08/12/2022 | R30954901 |
3469480 | Certificate of Analysis | 06/08/2022 | R30954901 |
3312586 | Certificate of Analysis | 08/12/2021 | R30954901 |