Learn More
Memmert™ Reinforced Stainless Steel Shelf
Additional reinforced stainless steel shelf for reinforced chamber option
Brand:  Memmert™ IPP1060ecoB32549
 View more versions of this product
Product Code. 17170045
  • 745.00€ / Each
Estimated Shipment: 15-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • With guide bars and fixing screws
  • Requires option K1
  • Consider max. loading of chamber; max. loading 60 kg
Specifications
Specifications
IPP1060eco
Stainless Steel
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Reinforced Stainless Steel Shelf > IPP1060eco