Learn More
KGW Isotherm™ Refill for 1500mL Borosilicate Glass Cylindrical Dewar Container
Brand: KGW Isotherm™ 10114
This item is not returnable.
View return policy
Specifications
115mm | |
245mm | |
1500 mL | |
Borosilicate Glass |
Cooling of small samples, Storage: liquid nitrogen ( LN2 ), dry ice ( CO2 )other coolants | |
300mm | |
90 mm |
Product Content Correction
Product Title
KGW Isotherm™ Refill for 1500mL Borosilicate Glass Cylindrical Dewar Container > I.D.: 90mm; O.D.: 115mm; Inner Height: 245mm; Outer Height: 300mm