Learn More
SI Analytics™ Reference Electrode
SI Analytics™ Reference Electrode is a low maintenance electrode with a screw plug head.
Brand: SI Analytics™ 285111767
Description
- Ag / AgCl reference
- PVDF diaphragm
- Paste electrolyte
Specifications
Screw Plug Head | |
B 1881 HD | |
120 mm |
PVDF Shaft | |
-5°C to +100°C | |
12 mm |