missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VEGF-120 Protein
Recombinant Protein
112.00€ - 5600.00€
Specifications
Accession Number | NP_001273987.1 |
---|---|
Name | VEGF-120 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Gene Symbol | VEGFA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15935709
|
enQuireBio™
QP5215-2ug |
2 μg |
112.00€
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15915709
|
enQuireBio™
QP5215-10ug |
10 μg |
263.00€
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15905709
|
enQuireBio™
QP5215-100ug |
100 μg |
730.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15945709
|
enQuireBio™
QP5215-500ug |
500 μg |
3430.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15925709
|
enQuireBio™
QP5215-1mg |
1 mg |
5600.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_001273987.1 | |
Research Use Only | |
VEGFA | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
VEGF-120 Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKCDKPR R | |
Greater than 95% as determined by SDS-PAGE. |