missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse VEGF-120 Protein
Recombinant Protein
Brand: enQuireBio™ QP5215-1mg
This item is not returnable.
View return policy
Specifications
NP_001273987.1 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MAPTTEGEQK SHEVIKFMDV YQRSYCRPIE TLVDIFQEYP DEIEYIFKPS CVPLMRCAGC CNDEALECVP TSESNITMQI MRIKPHQSQH IGEMSFLQHS RCECRPKKDR TKPEKCDKPR R | |
Greater than 95% as determined by SDS-PAGE. |
VEGF-120 Protein | |
Research Use Only | |
VEGFA | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |