Learn More
enQuireBio™ Recombinant Mouse Retnla Protein
Recombinant Protein
Brand:  enQuireBio™ QP5432-500ug
 View more versions of this product
Product Code. 15926119
  • 2710.00€ / 500µg
Explore more special offers
Specifications
NP_065255.2
Retnla Protein
Research Use Only
Retnla
Mouse
Untagged
10 mM sodium phosphate, pH 7.5
57262
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS
Greater than 95% as determined by SDS-PAGE.