missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Retnla Protein
Recombinant Protein
116.00€ - 4410.00€
Specifications
Accession Number | NP_065255.2 |
---|---|
Gene ID (Entrez) | 57262 |
Name | Retnla Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15936119
|
enQuireBio™
QP5432-5ug |
5 μg |
116.00€
5µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15916119
|
enQuireBio™
QP5432-25ug |
25 μg |
273.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15996109
|
enQuireBio™
QP5432-100ug |
100 μg |
682.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15926119
|
enQuireBio™
QP5432-500ug |
500 μg |
2710.00€
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15906119
|
enQuireBio™
QP5432-1mg |
1 mg |
4410.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_065255.2 | |
Retnla Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MDETIEIIVE NKVKELLANP ANYPSTVTKT LSCTSVKTMN RWASCPAGMT ATGCACGFAC GSWEIQSGDT CNCLCLLVDW TTARCCQLS | |
Greater than 95% as determined by SDS-PAGE. |
57262 | |
Research Use Only | |
Retnla | |
Mouse | |
Untagged | |
10 mM sodium phosphate, pH 7.5 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title