missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Sorting nexin-20 Protein
A DNA sequence encoding the Homo sapiens (Human) Sorting nexin-20, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6709-ec-10ug
This item is not returnable.
View return policy
Specifications
NP_001138444.1 | |
Sorting nexin-20 Protein | |
Research Use Only | |
SNX20 | |
Recombinant Protein | |
E. coli | |
MASPEHPGSP GCMGPITQCT ARTQQEAPAT GPDLPHPGPD GHLDTHSGLS SNSSMTTREL QQYWQNQKCR WKHVKLLFEI ASARIEERKV SKFVVYQIIV IQ | |
Greater than 90% as determined by SDS-PAGE. |
124460 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Sorting nexin-20 Protein > 10μg
Spot an opportunity for improvement?Share a Content Correction