missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Natriuretic peptides B Protein
A DNA sequence encoding the Homo sapiens (Human) Natriuretic peptides B, was expressed in the hosts and tags indicated.
160.00€ - 2485.00€
Specifications
Accession Number | NP_002512.1 |
---|---|
Gene ID (Entrez) | 4879 |
Name | Natriuretic peptides B Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16063250
|
enQuireBio™
QP6430-ec-10ug |
10 μg | N/A | ||||||
16003260
|
enQuireBio™
QP6430-ec-50ug |
50 μg | N/A | ||||||
16053250
|
enQuireBio™
QP6430-ec-100ug |
100 μg |
554.00€
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16083250
|
enQuireBio™
QP6430-ec-200ug |
200 μg | N/A | ||||||
16093250
|
enQuireBio™
QP6430-ec-500ug |
500 μg | N/A | ||||||
16073250
|
enQuireBio™
QP6430-ec-1mg |
1 mg |
2485.00€
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_002512.1 | |
Natriuretic peptides B Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
4879 | |
Research Use Only | |
NPPB | |
Recombinant Protein | |
E. coli | |
DLETSGLQEQ RNHLQGKLSE LQVEQTSLEP LQESP | |
Greater than 80% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title