missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Natriuretic peptides B Protein
A DNA sequence encoding the Homo sapiens (Human) Natriuretic peptides B, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6430-ec-1mg
This item is not returnable.
View return policy
Specifications
NP_002512.1 | |
Natriuretic peptides B Protein | |
Research Use Only | |
NPPB | |
Recombinant Protein | |
E. coli | |
DLETSGLQEQ RNHLQGKLSE LQVEQTSLEP LQESP | |
Greater than 80% as determined by SDS-PAGE. |
4879 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |