missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LGALS8 Protein

A cDNA sequence encoding the LGALS8 was constructed and used to recombinantly synthesize the protein.

137.00€ - 6815.00€

Specifications

Name LGALS8 Protein
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Biological Activity The ED50 of Galectin-8 as determined by its ability to agglutinate human red blood cells is 0.8∼4 microg/ml.
Product Type Recombinant Protein
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15945108
View Documents Promotion Details
enQuireBio™
QP10768-2ug
2 μg
137.00€
2µg
Please to purchase this item. Need a web account? Register with us today!
15925108
View Documents Promotion Details
enQuireBio™
QP10768-10ug
10 μg
212.00€
10µg
Please to purchase this item. Need a web account? Register with us today!
15935108
View Documents Promotion Details
enQuireBio™
QP10768-1mg
1 mg
6815.00€
1mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

LGALS8 Protein
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MMLSLNNLQN IIYNPVIPFV GTIPDQLDPG TLIVIRGHVP SDADRFQVDL QNGSSMKPRA DVAFHFNPRF KRAGCIVCNT LINEKWGREE ITYDTPFKRE KSFEIVIMVL KDKFQVAVNG KHTLLYGHRI GPEKIDTLGI YGKVNIHSIG FSFSSDLQST QASSLELTEI SRENVPKSGT PQLRLPFAAR LNTPMGPGRT VVVKGEVNAN AKSFNVDLLA GKSKDIALHL NPRLNIKAFV RNSFLQESWG EEERNITSFP FSPGMYFEMI IYCDVREFKV AVNGVHSLEY KHRFKELSSI DTLEINGDIH LLEVRSW
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
The ED50 of Galectin-8 as determined by its ability to agglutinate human red blood cells is 0.8∼4 microg/ml.
Human
Untagged
LGALS8 was lyophilized from a concentrated (1 mg/ml) solution in 20mM PBS, pH 7.4.
Videos
SDS
Documents

Documents

Product Certifications