missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human LGALS8 Protein
A cDNA sequence encoding the LGALS8 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10768-1mg
This item is not returnable.
View return policy
Specifications
LGALS8 Protein | |
Research Use Only | |
The ED50 of Galectin-8 as determined by its ability to agglutinate human red blood cells is 0.8∼4 microg/ml. | |
Human | |
Untagged | |
LGALS8 was lyophilized from a concentrated (1 mg/ml) solution in 20mM PBS, pH 7.4. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MMLSLNNLQN IIYNPVIPFV GTIPDQLDPG TLIVIRGHVP SDADRFQVDL QNGSSMKPRA DVAFHFNPRF KRAGCIVCNT LINEKWGREE ITYDTPFKRE KSFEIVIMVL KDKFQVAVNG KHTLLYGHRI GPEKIDTLGI YGKVNIHSIG FSFSSDLQST QASSLELTEI SRENVPKSGT PQLRLPFAAR LNTPMGPGRT VVVKGEVNAN AKSFNVDLLA GKSKDIALHL NPRLNIKAFV RNSFLQESWG EEERNITSFP FSPGMYFEMI IYCDVREFKV AVNGVHSLEY KHRFKELSSI DTLEINGDIH LLEVRSW | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |