missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human FGF-10 Protein

Recombinant Protein

119.00€ - 2585.00€

Specifications

Accession Number O15520, O15520, O15520
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 2 x PBS, pH 7.4.
Gene ID (Entrez) 2255
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18403752
View Documents
Novus Biologicals™
NBP2-34927-1mg
1 mg
2585.00€
1mg
Please to purchase this item. Need a web account? Register with us today!
18493802
View Documents
Novus Biologicals™
NBP2-34927-100ug
100 μg
852.00€
100µg
Please to purchase this item. Need a web account? Register with us today!
18494002
View Documents
Novus Biologicals™
NBP2-34927-5ug
5 μg
119.00€
5µg
Please to purchase this item. Need a web account? Register with us today!
18480522
View Documents
Novus Biologicals™
NBP2-34927-500ug
500 μg
1860.00€
500µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fully biologically active when compared to standard. The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 ng/mL, corresponding to a specific activity of >2.0 x 106IU/mg.
Specifications

Specifications

O15520, O15520, O15520
PAGE, Bioactivity
2255
Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
FGF-10, fibroblast growth factor 10, Keratinocyte growth factor 2, KGF-2/FGF-10, produced by fibroblasts of urinary bladder lamina propria
Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
>97% pure by SDS-PAGE and HPLC
LYOPH
Lyophilized from a 0.2 um filtered concentrated solution in 2 x PBS, pH 7.4.
19.1 kDa
Less than 1 EU/ug of FGF-10 as determined by LAL method.
FGF10
Angiogenesis, Biologically Active Proteins, Cell Cycle and Replication
FGF-10
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Novus Biologicals™ Recombinant Human FGF-10 Protein

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.