missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Canine IL-21 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
447.00€
Specifications
Gene ID (Entrez) | 442935 |
---|---|
Molecular Weight (g/mol) | 15.0 kDa |
Name | IL-21 |
Quantity | 25 μg |
Source | E. coli-derived canine IL-21 protein His18-Ser146,with an N-terminal Met |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18116490
|
R&D Systems™
5849-ML-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
447.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18363748
|
R&D Systems™
5849-ML-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
447.00€
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Canine IL-21 Protein is derived from E. coli. The Recombinant Canine IL-21 Protein has been validated for the following applications: Bioactivity.Specifications
442935 | |
IL-21 | |
E. coli-derived canine IL-21 protein His18-Ser146,with an N-terminal Met | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
15.0 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Recombinant Canine IL-21 Protein