missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rabbit anti-Syrian Hamster IgG (H+L) Secondary Antibody, AP
Rabbit Polyclonal Secondary Antibody
Brand: Invitrogen™ PA130348
This item is not returnable.
View return policy
Description
PA1-30348 detects Hamster IgG from hamster samples.
Anti-Hamster secondary antibodies are affinity-purified antibodies with well-characterized specificity for hamster immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Syrian Hamster IgG (H+L) | |
Polyclonal | |
Alkaline Phosphatase (AP) | |
Rabbit | |
IgG fraction | |
RUO | |
Hamster | |
Secondary Antibody | |
IgG |
Immunohistochemistry, ELISA, Western Blot | |
1 mg/mL | |
0.05M tris with 10mg/mL BSA, 50% glycerol, 0.001M MgCl2, 0.15M NaCl, 0.0001M ZnCl2 and 0.01% sodium azide; pH 8.0 | |
hamster IgG whole molecule. | |
1 mg | |
Secondary | |
4°C, do not freeze | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Rabbit anti-Syrian Hamster IgG (H+L) Secondary Antibody, AP > 1 mg; Alkaline Phosphatase (AP)
Spot an opportunity for improvement?Share a Content Correction