missing translation for 'onlineSavingsMsg'
Learn More
(R)-(-)-Prolinol, 98+%, Thermo Scientific Chemicals Product Code.: 15408939

(R)-(-)-Prolinol, 98+%, Thermo Scientific Chemicals

Product Code. 15408939
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 15408939

Brand: Thermo Scientific Alfa Aesar L11109.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68832-13-3
Molecular Formula C5H11NO
Molecular Weight (g/mol) 101.149
MDL Number MFCD00064321
InChI Key HVVNJUAVDAZWCB-RXMQYKEDSA-N
Synonym d-prolinol, r-pyrrolidin-2-ylmethanol, 2r-pyrrolidin-2-ylmethanol, r---2-pyrrolidinemethanol, r-prolinol, 2r-pyrrolidin-2-yl methanol, r-1-pyrrolidin-2-yl-methanol, 2-pyrrolidinemethanol, 2r, r---2-hydroxymethyl pyrrolidine
PubChem CID 2724541
ChEBI CHEBI:84258
IUPAC Name [(2R)-pyrrolidin-2-yl]methanol
SMILES C1CC(NC1)CO

Specifications

Refractive Index 1.4849
Density 1.025
Boiling Point 74°C to 76°C (2mmHg)
Quantity 25 g
Flash Point 86°C (186°F)
Optical Rotation −31° (c=1 in Toluene)
Sensitivity Air sensitive; Hygroscopic
Beilstein 1523669
Formula Weight 101.15
Percent Purity ≥98%
Chemical Name or Material (R)-(-)-Prolinol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10122519 Certificate TypeCertificate of Analysis Date16/06/2023 Catalog NumberL11109,L11109.03,L11109.14,L11109.06
Lot Number10122519 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL11109.03,L11109.14,L11109.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-(-)-Prolinol, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.