Learn More
R&D Systems™ Mouse LPAR3/LPA3/EDG-7 (NP_075359) VersaClone cDNA
Lysophosphatidic Acid Receptor 3
Brand:  R&D Systems™ RDC0561
Product Code. 18452280
  • 322.00€ / 10µg
Explore more special offers
Description
Description
The Mouse LPAR3/LPA3/EDG-7 (NP_075359) VersaClone cDNA clone contains the complete ORF for LPAR3/LPA3/EDG-7, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The LPAR3/LPA3/EDG-7 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Order Info
Ships at ambient temperature.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
mLPAR3
10 μg at 0.2 μg/μL
1078bp
Mouse
NP_075359
cDNA is provided in 10mM Tris-Cl, pH 8.5
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
R&D Systems™ Mouse LPAR3/LPA3/EDG-7 (NP_075359) VersaClone cDNA > 10ug
For Reasearch Use Only; Not for Use in Humans