Learn More
(R)-(-)-2-Amino-1-butanol, 98%, Thermo Scientific Chemicals Product Code.: 11440708
(R)-(-)-2-Amino-1-butanol, 98%, Thermo Scientific Chemicals
Product Code. 11440708
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
Product Code. 11440708
Brand: Thermo Scientific Alfa Aesar B25212.14
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 5856-63-3
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00064419
InChI Key JCBPETKZIGVZRE-UHFFFAOYNA-N
Synonym r-2-aminobutan-1-ol, r---2-amino-1-butanol, 2r-2-aminobutan-1-ol, r-2-amino-1-butanol, 1-butanol, 2-amino-, 2r, unii-4f725v55ln, 2-amino-1-butanol, r, 2-amino-1-butanol,-, l---2-amino-1-butanol, 1-butanol, 2-amino-, rShow MoreShow Less
PubChem CID 2723856
IUPAC Name 2-aminobutan-1-ol
SMILES CCC(N)CO
Specifications
Density 0.947
pH 11.1
Boiling Point 176°C to 178°C
Flash Point 89°C (192°F)
Odor Amine-like
Refractive Index 1.4525
Quantity 25 g
UN Number UN2735
Beilstein 1718929
Sensitivity Air sensitive; Hygroscopic
Optical Rotation −10° (Neat)
Formula Weight 89.14
Percent Purity 98%
Chemical Name or Material (R)-(-)-2-Amino-1-butanol
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ7889A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB25212.06,B25212.22,B25212.14
RUO – Research Use Only
Product Content Correction
Product Title
(R)-(-)-2-Amino-1-butanol, 98%, Thermo Scientific Chemicals >