Learn More
Contec™ Quiltec™ I Mop Head
Contec Quiltec I Mop Heads are manufactured from 100% 2-ply polyester around a rayon core so are ideal for even the highest grade of cleanroom as they generate very small numbers of particle and fibres.
197.00€ - 204.00€
Specifications
Color Code | White |
---|---|
Product Type | Mop Head |
Autoclavable | Not Autoclavable |
Disposable | Single-use |
Color-coded | No |
Product Code | Brand | Sterility | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Price | Quantity & Availability | ||||||
15815597
![]() |
Contec™
MHQT0100 |
Non-sterile |
197.00€
Pack of 40 |
In Stock
Log in to see stock. |
||||||
15817044
![]() |
Contec™
MHQT0150 |
Sterile |
204.00€
Pack of 40 |
Estimated Shipment: 11-06-2025
Log in to see stock. |
||||||
Description
- Two-ply 100% knitted polyester over a rayon core
- Gamma irradiated at no less than 25 kGy
- 5 mops double bagged
- Approx 18-20 sqm coverage
- Certificates of conformity supplied with every batch
Specifications
White | |
Not Autoclavable | |
No |
Mop Head | |
Single-use |
Product Title
Contec™ Quiltec™ I Mop Head