Learn More
Quickfit™ Screwthread Adapter to Fit Cone
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
Brand:  Quickfit ST52/18
39.85 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 10654969
  • 48.30€ / Pack of 2
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Description
Secure thermometers and tubes to glass containers. The QuickFit™ screwthread adapter comes in a variety of sizes to fit a variety of container and inlets.
  • With screw thread joint complete with plastic cap, silicone rubber ring and PTFE washer
  • Ideal for securing thermometers, gas inlet tubes, etc
TRUSTED_SUSTAINABILITY
Specifications
Specifications
19/26
Plastic cap, Silicone Rubber ring and PTFE washer
Cone screw thread adapter
Ideal for securing thermometers, gas inlet tubes
Borosilicate Glass
18
Documents
Product Information
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    28
    Hours
    7
    Minutes
    35
    Seconds
    59
Product Content Correction
Product Title
Quickfit™ Screwthread Adapter to Fit Cone > Dia. through cap: 7 to 8.5mm; Cone size: 19/26; Thread size: 18