Learn More
Quickfit™ Funnel with Ground Glass Cone
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Quickfit™ Funnel with Ground Glass Cone is designed for transferring powders into Quickfit™ flasks and vessels.
Brand:  Quickfit CF5/3
46.44 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 12438816
  • 54.05€ / Each
Estimated Shipment: 10-06-2025
to see stock.
Explore more special offers
Description
Manufactured from chemically resistant Pyrex™ Borosilicate Glass. Quickfit™ Funnel with Ground Glass Cone is designed for transferring powders into Quickfit™ flasks and vessels.
  • Designed for transferring powders into Quickfit™ flasks and vessels
  • With Quickfit™ ground glass cone
  • Manufactured from chemically resistant Pyrex™ Borosilicate Glass
TRUSTED_SUSTAINABILITY
Specifications
Specifications
105mm
24/29
Powder Funnel
Borosilicate Glass
Quickfit™ Ground Glass Cone
Promotions
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    22
    Hours
    15
    Minutes
    39
    Seconds
    59
Product Content Correction
Product Title
Quickfit™ Funnel with Ground Glass Cone > Cone Size: 24/29