Learn More
Hellma™ Quartz Flow Through Cells Far UV
Brand: Hellma™ 170-700-1-40
This item is not returnable.
View return policy
Specifications
Flow-through Cell Far UV | |
Quartz | |
1 mm |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Hellma™ Quartz Flow Through Cells Far UV > Light Path: 1mm