Learn More
Applied Biosystems™ QuantStudio 6 Flex package
Facilitates real-time PCR based applications with an upgradable block selection. With a simplified workflow, intuitive software, and touch-screen interface, the QuantStudio™ 6 Flex system offers exceptional reproducibility at a price you can afford.
Brand:  Applied Biosystems™ A27179
 View more versions of this product
Product Code. 15761168
  • 82656.00€ / Each
Explore more special offers
Description
Description
  • Affordable price allows you to obtain a high performance instrument with minimal upfront capital investment
  • Compatible with over 8 million TaqMan™ assays for gene expression, genetic variation, gene regulation, or protein expression experiments
  • The intelligent design of the QuantStudio™ platform combined with Applied Biosystems™ instrument reliability and accuracy
  • Easy-to-use software, responsive touch-screen, and effortless block exchange without the need for any tools
  • The QuantStudio™ 6 Flex system accommodates the interchange of a 96-well, 96-well Fast, or 384-well block
  • The QuantStudio™ 6 Flex system can also be upgraded to a QuantStudio™ 7 Flex system at a later date, as your needs change
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PC (Laptop)
96
54.7 cm
1 Year Manufacturers + 1 Year Field Service repair (with 1 Planned Maintenance)
4°C to 99.9°C
96-well FAST, 6.5°C/s
Excitation 455 to 672 nm, Emission 505 to 723 nm
FAM™, SYBR™ Green, VIC™, JOE™, HEX™, TET™, ABY™, NED™, TAMRA™, Cy™3, JUN™, ROX™, Texas Red™, Mustang Purple™, Cy™5, LIZ™, Cy™5.5
96-well FAST
0.25°C
Touchscreen
33.8 cm
52.5 cm
QuantStudio 6 Flex
6.0°C/s (96-well FAST)
0.1 ml (96-well FAST)
38 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Applied Biosystems™ QuantStudio 6 Flex package > PC: Laptop, Block: 96-well FAST, Warranty: 1 year manufacturers + 1 year Field Service repair (with 1 planned maintenance)