Learn More
Invitrogen™ Quant-iT™ Endotoxin Detection Assay Kit
The Invitrogen Quant-iT Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on fluorescence microplate readers that uses amebocyte lysates to quantify endotoxin in protein, peptide, antibody, or nucleic acid samples.
Brand:  Invitrogen™ Q32892
548.72 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Product Code. 17942031
  • 760.00€ / 160 reactions
Explore more special offers
Description
Description
The Invitrogen Quant-iT Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on fluorescence microplate readers that uses amebocyte lysates to quantify endotoxin in protein, peptide, antibody, or nucleic acid samples. Amebocyte lysates are widely used as a simple and sensitive assay for the detection of endotoxin lipopolysaccharide (LPS), the membrane component of gram-negative bacteria.
Features of the Quant-iT Endotoxin Detection Assay Kit include:
Highly sensitive with a broad range—detect as little as 0.01 EU/mL to 10.0 EU/mL
Specific—no interference from ß-glucans and suitable for wide range of samples, including protein, vaccine, plasmid, DNA, and RNA
Easy to use—the simple workflow requires one incubation prior to measurement
Endotoxin levels in the samples are accurately determined using the included endotoxin standard of known concentration that is derived from E. coli strain O111: B4. Determining endotoxin levels is important to assess the efficiency of endotoxin removal methods and prevent endotoxic shock, inflammation, and/or sepsis in tissue culture cells and animals injected with endotoxin-contaminated proteins.
When endotoxin encounters the amebocyte lysate, a series of enzymatic reactions results in the activation of Factor C, Factor B, and pro-clotting enzyme. The activated enzyme catalyzes a cleavage event in the substrate to produce a strong fluorescent signal. After stopping the reaction, the resulting signal is measured on a fluorescence microplate reader (such as Varioskan LUX). The correlation between fluorescent signal is proportional to the endotoxin concentration in the sample.
Notes
• Compatible with Pierce Endotoxin-Free Plate Strips (Cat. No. A43888)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Endotoxin
Kit
160 reactions
Protein, Antibody, Plasmid, DNA, RNA
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2978481 Certificate TypeCertificate of Analysis Date26/08/2024 Catalog NumberQ32892
Lot Number2916404 Certificate TypeCertificate of Analysis Date30/05/2024 Catalog NumberQ32892
Lot Number2896645 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog NumberQ32892
Lot Number2811999 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog NumberQ32892
Lot Number2734935 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog NumberQ32892
Product Title
Invitrogen™ Quant-iT™ Endotoxin Detection Assay Kit > 160 Reactions