Learn More
Pyrex™ Heavy Walled Test Tube
Pyrex™ Heavy Walled Test Tube is designed to provide extra mechanical strength.
Brand:  Pyrex 1626/13M
198.45 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 15566745
  • 216.00€ / Pack of 100
Estimated Shipment: 13-08-2025
to see stock.
Explore more special offers
Description
  • Conforms to ISO 4142
  • Rimless test tubes with a heavy wall to provide extra mechanical strength
  • Robust construction reduces possibility of breakages: - Lower replacement costs - Improved user safety
  • Manufactured from Pyrex™ borosilicate glass for high resistance to thermal shock and chemical attack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass
100/Pk.
Test Tube
Promotions
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    33
    Hours
    21
    Minutes
    44
    Seconds
    5
Product Content Correction
Product Title
Pyrex™ Heavy Walled Test Tube > Quantity: 100/Pk