Learn More
Pyrex™ Dessicator Plates
For use with Pyrex vacuum and non-vacuum desiccators
Brand: Pyrex 1597/02D
This item is not returnable.
View return policy
Description
For use with Pyrex vacuum and non-vacuum desiccators
- For use with Pyrex vacuum and non-vacuum desiccators
Specifications
Plate Size: 150 mm | |
Metal | |
Pyrex vacuum and non-vacuum desiccator |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Pyrex™ Dessicator Plates > Plate Size: 150mm