Learn More
Cole-Parmer PVC Lab and Vacuum Tubing
Smooth, polished inner wall prevents buildups—ideal for general laboratory applications
Brand:  Cole-Parmer 50121-16
 View more versions of this product
Product Code. 30053338
  • 32.75€ / 15m
Estimated Shipment: 12-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Versatile applications including peristaltic pumps, analytical instruments, incubators, gas, and drain lines. Glass-smooth inner bore design for easy clean-ups and changeovers, enhancing workflow efficiency. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Specifications
Specifications
1/16 in.
1/8 in.
Translucent
PVC
50 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Cole-Parmer PVC Lab and Vacuum Tubing > 1/16 in., 1/8 in.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.